Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606845 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCBLD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DCBLD1 - discoidin, CUB and LCCL domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173674.2 | 4455 | Intron | NP_775945.1 | |||
XM_005266941.1 | 4455 | Intron | XP_005266998.1 | |||
XM_006715463.1 | 4455 | Intron | XP_006715526.1 | |||
XM_011535771.1 | 4455 | Intron | XP_011534073.1 | |||
XM_011535773.2 | 4455 | Intron | XP_011534075.1 | |||
XM_011535774.2 | 4455 | Intron | XP_011534076.1 | |||
XM_017010775.1 | 4455 | Intron | XP_016866264.1 |
GOPC - golgi associated PDZ and coiled-coil motif containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017408.2 | 4455 | UTR 3 | NP_001017408.1 | |||
NM_020399.3 | 4455 | UTR 3 | NP_065132.1 |
Set Membership: |
HapMap Validated |