Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGCTGGTGGGAGTTGACGACG[C/T]GGTGCTGGGCGTTGGGACCCTACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604857 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRP54 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SRP54 - signal recognition particle 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146282.1 | 407 | UTR 5 | NP_001139754.1 | |||
NM_003136.3 | 407 | UTR 5 | NP_003127.1 | |||
XM_005268024.2 | 407 | UTR 5 | XP_005268081.1 | |||
XM_011537106.1 | 407 | Intron | XP_011535408.1 | |||
XM_017021615.1 | 407 | UTR 5 | XP_016877104.1 |
Set Membership: |
HapMap |