Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300415 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTM1 - myotubularin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000252.2 | Intron | NP_000243.1 | ||||
XM_005274687.2 | Intron | XP_005274744.1 | ||||
XM_011531171.1 | Intron | XP_011529473.1 | ||||
XM_011531172.1 | Intron | XP_011529474.1 | ||||
XM_011531173.2 | Intron | XP_011529475.1 | ||||
XM_017029547.1 | Intron | XP_016885036.1 | ||||
XM_017029548.1 | Intron | XP_016885037.1 | ||||
XM_017029549.1 | Intron | XP_016885038.1 | ||||
XM_017029550.1 | Intron | XP_016885039.1 | ||||
XM_017029551.1 | Intron | XP_016885040.1 |
Set Membership: |
HapMap |