Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607650 MIM: 615997 | ||||||||||||||||||||||||||||||||
Literature Links: |
DEFB118 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
DEFB118 - defensin beta 118 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DEFB119 - defensin beta 119 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271209.1 | 456 | Intron | NP_001258138.1 | |||
NM_153289.3 | 456 | UTR 3 | NP_695021.2 | |||
NM_153323.4 | 456 | Intron | NP_697018.1 |
Set Membership: |
HapMap |