Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608685 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIBC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RIBC2 - RIB43A domain with coiled-coils 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015653.4 | 53 | UTR 5 | NP_056468.3 | |||
XM_005261524.4 | 53 | Intron | XP_005261581.1 | |||
XM_011530126.2 | 53 | Intron | XP_011528428.1 | |||
XM_017028766.1 | 53 | Intron | XP_016884255.1 |
SMC1B - structural maintenance of chromosomes 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291501.1 | 53 | Intron | NP_001278430.1 | |||
NM_148674.4 | 53 | Intron | NP_683515.4 | |||
XM_011530144.2 | 53 | Intron | XP_011528446.1 | |||
XM_011530145.2 | 53 | Intron | XP_011528447.1 |
Set Membership: |
HapMap Validated |