Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604414 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf23 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C22orf23 - chromosome 22 open reading frame 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6820 - microRNA 6820 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2F - RNA polymerase II subunit F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301129.1 | Intron | NP_001288058.1 | ||||
NM_001301130.1 | Intron | NP_001288059.1 | ||||
NM_001301131.1 | Intron | NP_001288060.1 | ||||
NM_021974.4 | Intron | NP_068809.1 | ||||
XM_017028832.1 | Intron | XP_016884321.1 |
Set Membership: |
HapMap Validated |