Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605926 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PICK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PICK1 - protein interacting with PRKCA 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039583.1 | 142 | Intron | NP_001034672.1 | |||
NM_001039584.1 | 142 | Intron | NP_001034673.1 | |||
NM_012407.3 | 142 | Intron | NP_036539.1 | |||
XM_006724379.2 | 142 | Intron | XP_006724442.1 | |||
XM_006724380.2 | 142 | Intron | XP_006724443.1 | |||
XM_011530517.2 | 142 | Intron | XP_011528819.1 | |||
XM_017029090.1 | 142 | Intron | XP_016884579.1 | |||
XM_017029091.1 | 142 | UTR 5 | XP_016884580.1 | |||
XM_017029092.1 | 142 | Intron | XP_016884581.1 | |||
XM_017029093.1 | 142 | Intron | XP_016884582.1 | |||
XM_017029094.1 | 142 | UTR 5 | XP_016884583.1 | |||
XM_017029095.1 | 142 | UTR 5 | XP_016884584.1 |