Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGTTCCCTCCTCTTACGGACTGG[A/G]TTGGGATAGGGGTGGAGGCTCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138032 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLP1R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GLP1R - glucagon like peptide 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002062.4 | Intron | NP_002053.3 | ||||
XM_017010750.1 | Intron | XP_016866239.1 | ||||
XM_017010751.1 | Intron | XP_016866240.1 |
Set Membership: |
Validated |