Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGGCACTATGGTGGGGAAACC[A/G]GAGACAAGTAGCAAGTGAAATTTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608797 MIM: 601304 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf46 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C22orf46 - chromosome 22 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142964.1 | 1907 | UTR 3 | NP_001136436.1 | |||
XM_011530373.2 | 1907 | UTR 3 | XP_011528675.1 | |||
XM_017028939.1 | 1907 | UTR 5 | XP_016884428.1 |
MEI1 - meiotic double-stranded break formation protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNU13 - SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |