Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTTCACGCAGGATCTAACCTGT[A/G]GGTCTCACGTGCTTCCACCCCTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf195 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf195 - chromosome 1 open reading frame 195 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM51 - transmembrane protein 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136216.1 | Intron | NP_001129688.1 | ||||
NM_001136217.1 | Intron | NP_001129689.1 | ||||
NM_001136218.1 | Intron | NP_001129690.1 | ||||
NM_001319665.1 | Intron | NP_001306594.1 | ||||
NM_018022.2 | Intron | NP_060492.1 | ||||
XM_005245919.1 | Intron | XP_005245976.1 | ||||
XM_011541676.1 | Intron | XP_011539978.1 | ||||
XM_017001590.1 | Intron | XP_016857079.1 |
TMEM51-AS1 - TMEM51 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |