Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGTTTGTTGTTGTTGCTTTTG[A/G]AAAAATAGAGACAAGGTCTCTGTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf195 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf195 - chromosome 1 open reading frame 195 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM51 - transmembrane protein 51 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136216.1 | Intron | NP_001129688.1 | ||||
NM_001136217.1 | Intron | NP_001129689.1 | ||||
NM_001136218.1 | Intron | NP_001129690.1 | ||||
NM_001319665.1 | Intron | NP_001306594.1 | ||||
NM_018022.2 | Intron | NP_060492.1 | ||||
XM_005245919.1 | Intron | XP_005245976.1 | ||||
XM_011541676.1 | Intron | XP_011539978.1 | ||||
XM_017001590.1 | Intron | XP_016857079.1 |
TMEM51-AS1 - TMEM51 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |