Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTAAGCAGGTTGAGTTGGGTTTT[C/G]TATTTGCAATAGCAAAAGTCCTGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602049 MIM: 182453 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAC2 - ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002872.4 | 1454 | UTR 3 | NP_002863.1 | |||
XM_006724286.3 | 1454 | Intron | XP_006724349.1 |
SSTR3 - somatostatin receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |