Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGGGCTCCCTGTCCTTGTGGTA[C/T]ACTCATCTCACTGATTTTTAGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C6orf10 - chromosome 6 open reading frame 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286474.1 | 1149 | Missense Mutation | ATA,GTA | I,V 313 | NP_001273403.1 | |
NM_001286475.1 | 1149 | Missense Mutation | ATA,GTA | I,V 299 | NP_001273404.1 | |
NM_006781.4 | 1149 | Missense Mutation | ATA,GTA | I,V 315 | NP_006772.3 | |
XM_017010182.1 | 1149 | Missense Mutation | ATA,GTA | I,V 221 | XP_016865671.1 | |
XM_017010183.1 | 1149 | Intron | XP_016865672.1 | |||
XM_017010184.1 | 1149 | Intron | XP_016865673.1 | |||
XM_017010185.1 | 1149 | Missense Mutation | ATA,GTA | I,V 326 | XP_016865674.1 |
LOC101929163 - uncharacterized LOC101929163 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |