Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608544 MIM: 602399 MIM: 610053 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EUR
|
African American - Not Available | YRI (Yoruba)
|
||||||
EAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
||||||
SAS - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
||||||
AFR - Not Available | ||||||||
AMR
|
HDAC10 - histone deacetylase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159286.1 | Intron | NP_001152758.1 | ||||
NM_032019.5 | Intron | NP_114408.3 |
MAPK12 - mitogen-activated protein kinase 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBGCP6 - tubulin gamma complex associated protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |