Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154235 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAK - male germ cell associated kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242385.1 | 3428 | UTR 3 | NP_001229314.1 | |||
NM_001242957.2 | 3428 | UTR 3 | NP_001229886.1 | |||
NM_005906.5 | 3428 | UTR 3 | NP_005897.1 | |||
XM_011514619.2 | 3428 | UTR 3 | XP_011512921.1 | |||
XM_011514620.2 | 3428 | UTR 3 | XP_011512922.1 | |||
XM_011514621.2 | 3428 | Intron | XP_011512923.1 | |||
XM_011514622.2 | 3428 | UTR 3 | XP_011512924.1 | |||
XM_011514624.2 | 3428 | Intron | XP_011512926.1 | |||
XM_017010863.1 | 3428 | UTR 3 | XP_016866352.1 | |||
XM_017010864.1 | 3428 | UTR 3 | XP_016866353.1 | |||
XM_017010865.1 | 3428 | UTR 3 | XP_016866354.1 | |||
XM_017010866.1 | 3428 | Intron | XP_016866355.1 |
TMEM14B - transmembrane protein 14B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |