Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTTAGTTCTTACATGGCCCCCT[C/T]TCTCTTCTATGCTCAGATGTGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600713 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD11B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSD11B1 - hydroxysteroid 11-beta dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206741.1 | Intron | NP_001193670.1 | ||||
NM_005525.3 | Intron | NP_005516.1 | ||||
NM_181755.2 | Intron | NP_861420.1 |
LOC101930114 - uncharacterized LOC101930114 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |