Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACAAAAAACCCAATATAGAAAG[A/G]GGAGGTCCCAGAGATGCACCTAAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603978 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZSCAN12 - zinc finger and SCAN domain containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163391.1 | Intron | NP_001156863.1 | ||||
XM_011515014.2 | Intron | XP_011513316.1 | ||||
XM_011515015.2 | Intron | XP_011513317.1 | ||||
XM_011515017.2 | Intron | XP_011513319.1 | ||||
XM_017011528.1 | Intron | XP_016867017.1 |
Set Membership: |
HapMap |