Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZSCAN23 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZSCAN23 - zinc finger and SCAN domain containing 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012455.1 | 2900 | Intron | NP_001012458.1 | |||
XM_005248950.2 | 2900 | UTR 3 | XP_005249007.1 | |||
XM_011514406.2 | 2900 | Intron | XP_011512708.1 | |||
XM_011514408.2 | 2900 | Intron | XP_011512710.1 | |||
XM_011514410.2 | 2900 | Intron | XP_011512712.1 | |||
XM_017010479.1 | 2900 | Intron | XP_016865968.1 |
Set Membership: |
HapMap |