Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAACTAAAAGTAAGCTAGATCTG[A/T]TTCTATAATAACCTTACAAAACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615867 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TBC1D32 - TBC1 domain family member 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152730.5 | 4235 | UTR 3 | NP_689943.4 | |||
XM_005266861.2 | 4235 | UTR 3 | XP_005266918.1 | |||
XM_011535569.1 | 4235 | UTR 3 | XP_011533871.1 | |||
XM_011535570.1 | 4235 | UTR 3 | XP_011533872.1 | |||
XM_011535571.2 | 4235 | UTR 3 | XP_011533873.1 | |||
XM_011535574.1 | 4235 | UTR 3 | XP_011533876.1 | |||
XM_011535575.2 | 4235 | Intron | XP_011533877.1 | |||
XM_011535576.2 | 4235 | Intron | XP_011533878.1 | |||
XM_011535580.2 | 4235 | UTR 3 | XP_011533882.1 | |||
XM_011535582.2 | 4235 | Intron | XP_011533884.1 | |||
XM_011535585.2 | 4235 | Intron | XP_011533887.1 | |||
XM_017010397.1 | 4235 | UTR 3 | XP_016865886.1 | |||
XM_017010398.1 | 4235 | UTR 3 | XP_016865887.1 | |||
XM_017010399.1 | 4235 | UTR 3 | XP_016865888.1 | |||
XM_017010400.1 | 4235 | UTR 3 | XP_016865889.1 | |||
XM_017010401.1 | 4235 | UTR 3 | XP_016865890.1 | |||
XM_017010402.1 | 4235 | Intron | XP_016865891.1 | |||
XM_017010403.1 | 4235 | Intron | XP_016865892.1 | |||
XM_017010404.1 | 4235 | UTR 3 | XP_016865893.1 | |||
XM_017010405.1 | 4235 | Intron | XP_016865894.1 |