Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604009 MIM: 601661 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BAIAP3 - BAI1 associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199096.1 | Intron | NP_001186025.1 | ||||
NM_001199097.1 | Intron | NP_001186026.1 | ||||
NM_001199098.1 | Intron | NP_001186027.1 | ||||
NM_001199099.1 | Intron | NP_001186028.1 | ||||
NM_001286464.1 | Intron | NP_001273393.1 | ||||
NM_003933.4 | Intron | NP_003924.2 | ||||
XM_011522728.1 | Intron | XP_011521030.1 | ||||
XM_011522729.1 | Intron | XP_011521031.1 | ||||
XM_011522730.2 | Intron | XP_011521032.1 |
UBE2I - ubiquitin conjugating enzyme E2 I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |