Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604009 MIM: 607838 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAIAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
BAIAP3 - BAI1 associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNPTG - N-acetylglucosamine-1-phosphate transferase gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032520.4 | Intron | NP_115909.1 | ||||
XM_017023782.1 | Intron | XP_016879271.1 | ||||
XM_017023783.1 | Intron | XP_016879272.1 |
TSR3 - TSR3, acp transferase ribosome maturation factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNKL - unkempt family like zinc finger | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |