Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGATGGAGCATACCTCCAAACC[A/C]GTATACAATATAGACGGTGCCTCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606876 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3199-1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR3199-1 - microRNA 3199-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3199-2 - microRNA 3199-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNB - phosphatidylinositol transfer protein beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC28-AS1 - TTC28 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |