Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605860 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCAN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RCAN3 - RCAN family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251977.1 | Intron | NP_001238906.1 | ||||
NM_001251978.1 | Intron | NP_001238907.1 | ||||
NM_001251979.1 | Intron | NP_001238908.1 | ||||
NM_001251980.1 | Intron | NP_001238909.1 | ||||
NM_001251981.1 | Intron | NP_001238910.1 | ||||
NM_001251982.1 | Intron | NP_001238911.1 | ||||
NM_001251983.1 | Intron | NP_001238912.1 | ||||
NM_001251984.1 | Intron | NP_001238913.1 | ||||
NM_001251985.1 | Intron | NP_001238914.1 | ||||
NM_013441.3 | Intron | NP_038469.1 |
Set Membership: |
HapMap |