Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCAGGGGCCCATGACCAAGAAG[A/C]AAGCAAGCACCATGACTGGGTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600863 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1E PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CSNK1E - casein kinase 1 epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001894.4 | Intron | NP_001885.1 | ||||
NM_152221.2 | Intron | NP_689407.1 |
LOC400927-CSNK1E - LOC400927-CSNK1E readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289912.1 | Intron | NP_001276841.1 |
Set Membership: |
HapMap Validated |