Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131242 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EDN3 - endothelin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302455.1 | Intron | NP_001289384.1 | ||||
NM_001302456.1 | Intron | NP_001289385.1 | ||||
NM_207032.2 | Intron | NP_996915.1 | ||||
NM_207033.2 | Intron | NP_996916.1 | ||||
NM_207034.2 | Intron | NP_996917.1 | ||||
XM_005260312.4 | Intron | XP_005260369.1 | ||||
XM_005260313.4 | Intron | XP_005260370.1 | ||||
XM_006723734.3 | Intron | XP_006723797.1 | ||||
XM_011528655.2 | Intron | XP_011526957.1 |
Set Membership: |
HapMap Validated |