Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPPA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PAPPA2 - pappalysin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020318.2 | Intron | NP_064714.2 | ||||
NM_021936.2 | Intron | NP_068755.2 | ||||
XM_005245422.3 | Intron | XP_005245479.1 | ||||
XM_011509857.1 | Intron | XP_011508159.1 | ||||
XM_011509858.2 | Intron | XP_011508160.1 | ||||
XM_017002023.1 | Intron | XP_016857512.1 | ||||
XM_017002024.1 | Intron | XP_016857513.1 | ||||
XM_017002025.1 | Intron | XP_016857514.1 |