Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604971 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDFI PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MDFI - MyoD family inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300804.1 | Intron | NP_001287733.1 | ||||
NM_001300805.1 | Intron | NP_001287734.1 | ||||
NM_001300806.1 | Intron | NP_001287735.1 | ||||
NM_005586.3 | Intron | NP_005577.1 | ||||
XM_005249117.3 | Intron | XP_005249174.1 | ||||
XM_011514625.2 | Intron | XP_011512927.1 | ||||
XM_011514626.2 | Intron | XP_011512928.1 | ||||
XM_017010867.1 | Intron | XP_016866356.1 |
Set Membership: |
HapMap Validated |