Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCCTACGTTGTTCAGGGAATTG[C/T]GGAGGATACAGGATAACACATGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 111700 MIM: 610301 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHCE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RHCE - Rh blood group CcEe antigens | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM57 - transmembrane protein 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282564.1 | Intron | NP_001269493.1 | ||||
NM_018202.5 | Intron | NP_060672.2 | ||||
XM_005245931.2 | Intron | XP_005245988.1 | ||||
XM_011541704.2 | Intron | XP_011540006.1 |