Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTGTAGTGCTGTGTGAAGCTTA[A/G]GCAGTGCAGTATTGTACCTGACTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 111700 MIM: 610301 | ||||||||||||||||||||
Literature Links: |
RHCE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHCE - Rh blood group CcEe antigens | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM57 - transmembrane protein 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282564.1 | Intron | NP_001269493.1 | ||||
NM_018202.5 | Intron | NP_060672.2 | ||||
XM_005245931.2 | Intron | XP_005245988.1 | ||||
XM_011541704.2 | Intron | XP_011540006.1 |