Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTGGATTTGAATTATGGATCTG[A/G]TACTCTAATGGGCAAGGTGGCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615678 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3BGRL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SH3BGRL2 - SH3 domain binding glutamate rich protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031469.3 | Intron | NP_113657.1 | ||||
XM_011536165.2 | Intron | XP_011534467.1 | ||||
XM_011536282.2 | Intron | XP_011534584.1 | ||||
XM_017011344.1 | Intron | XP_016866833.1 | ||||
XM_017011345.1 | Intron | XP_016866834.1 |