Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGGTTGACAGTTATTGTAAAA[C/T]ATACCTTACTCCCATTCTGTCTTAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607820 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOOK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HOOK1 - hook microtubule-tethering protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015888.4 | Intron | NP_056972.1 | ||||
XM_006710676.1 | Intron | XP_006710739.1 | ||||
XM_011541562.2 | Intron | XP_011539864.1 | ||||
XM_011541563.1 | Intron | XP_011539865.1 | ||||
XM_017001424.1 | Intron | XP_016856913.1 |
Set Membership: |
HapMap |