Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607757 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBY1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CBY1 - chibby family member 1, beta catenin antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002880.1 | Intron | NP_001002880.2 | ||||
NM_015373.3 | Intron | NP_056188.1 |
FAM227A - family with sequence similarity 227 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013647.1 | Intron | NP_001013669.1 | ||||
NM_001291030.1 | Intron | NP_001277959.1 | ||||
XM_006724291.2 | Intron | XP_006724354.2 | ||||
XM_006724292.2 | Intron | XP_006724355.2 | ||||
XM_006724293.2 | Intron | XP_006724356.1 | ||||
XM_006724294.2 | Intron | XP_006724357.2 | ||||
XM_006724295.2 | Intron | XP_006724358.2 | ||||
XM_011530308.2 | Intron | XP_011528610.1 | ||||
XM_011530309.2 | Intron | XP_011528611.1 | ||||
XM_011530310.1 | Intron | XP_011528612.1 | ||||
XM_011530311.1 | Intron | XP_011528613.1 | ||||
XM_011530312.1 | Intron | XP_011528614.1 | ||||
XM_011530313.2 | Intron | XP_011528615.1 | ||||
XM_011530314.1 | Intron | XP_011528616.1 |