Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCGGATTTAATACAGTGTTAAT[C/T]ATAATAGCATGATTAATCATAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611690 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRRG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PRRG4 - proline rich and Gla domain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024081.5 | Intron | NP_076986.1 | ||||
XM_006718313.3 | Intron | XP_006718376.3 | ||||
XM_006718314.3 | Intron | XP_006718377.1 | ||||
XM_017018275.1 | Intron | XP_016873764.1 |
Set Membership: |
HapMap Validated |