Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617001 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENPP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ENPP5 - ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290072.1 | 2845 | UTR 3 | NP_001277001.1 | |||
NM_001290073.1 | 2845 | UTR 3 | NP_001277002.1 | |||
NM_021572.5 | 2845 | UTR 3 | NP_067547.1 | |||
XM_005249260.3 | 2845 | UTR 3 | XP_005249317.1 | |||
XM_011514786.2 | 2845 | UTR 3 | XP_011513088.1 |