Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603952 MIM: 607445 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DRG1 - developmentally regulated GTP binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF4ENIF1 - eukaryotic translation initiation factor 4E nuclear import factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164501.1 | Intron | NP_001157973.1 | ||||
NM_001164502.1 | Intron | NP_001157974.1 | ||||
NM_019843.3 | Intron | NP_062817.2 | ||||
XM_005261686.2 | Intron | XP_005261743.1 | ||||
XM_005261687.2 | Intron | XP_005261744.1 | ||||
XM_005261688.2 | Intron | XP_005261745.1 | ||||
XM_006724281.3 | Intron | XP_006724344.3 | ||||
XM_011530280.1 | Intron | XP_011528582.1 | ||||
XM_011530281.2 | Intron | XP_011528583.1 | ||||
XM_017028863.1 | Intron | XP_016884352.1 | ||||
XM_017028864.1 | Intron | XP_016884353.1 |
Set Membership: |
HapMap |