Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607379 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NF2 - neurofibromin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000268.3 | 240 | UTR 5 | NP_000259.1 | |||
NM_016418.5 | 240 | UTR 5 | NP_057502.2 | |||
NM_181825.2 | 240 | UTR 5 | NP_861546.1 | |||
NM_181828.2 | 240 | UTR 5 | NP_861966.1 | |||
NM_181829.2 | 240 | UTR 5 | NP_861967.1 | |||
NM_181830.2 | 240 | UTR 5 | NP_861968.1 | |||
NM_181831.2 | 240 | UTR 5 | NP_861969.1 | |||
NM_181832.2 | 240 | UTR 5 | NP_861970.1 | |||
NM_181833.2 | 240 | UTR 5 | NP_861971.1 | |||
XM_017028809.1 | 240 | Intron | XP_016884298.1 | |||
XM_017028810.1 | 240 | Intron | XP_016884299.1 |