Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607379 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NF2 - neurofibromin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000268.3 | Intron | NP_000259.1 | ||||
NM_016418.5 | Intron | NP_057502.2 | ||||
NM_181825.2 | Intron | NP_861546.1 | ||||
NM_181828.2 | Intron | NP_861966.1 | ||||
NM_181829.2 | Intron | NP_861967.1 | ||||
NM_181830.2 | Intron | NP_861968.1 | ||||
NM_181831.2 | Intron | NP_861969.1 | ||||
NM_181832.2 | Intron | NP_861970.1 | ||||
NM_181833.2 | Intron | NP_861971.1 | ||||
XM_017028809.1 | Intron | XP_016884298.1 | ||||
XM_017028810.1 | Intron | XP_016884299.1 |
Set Membership: |
HapMap |