Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCATGACTTCAGTATTAAGAACC[A/G]TAGCGATCCTGAAGAGAGAACCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612439 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARFGAP3 - ADP ribosylation factor GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142293.1 | 1762 | Silent Mutation | TAC,TAT | Y,Y 470 | NP_001135765.1 | |
NM_014570.4 | 1762 | Silent Mutation | TAC,TAT | Y,Y 514 | NP_055385.3 | |
XM_005261525.3 | 1762 | Silent Mutation | TAC,TAT | Y,Y 470 | XP_005261582.1 |
Set Membership: |
HapMap |