Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605942 MIM: 604714 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DSE - dermatan sulfate epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080976.2 | 4802 | Intron | NP_001074445.1 | |||
NM_001322937.1 | 4802 | Intron | NP_001309866.1 | |||
NM_001322938.1 | 4802 | Intron | NP_001309867.1 | |||
NM_001322939.1 | 4802 | Intron | NP_001309868.1 | |||
NM_001322940.1 | 4802 | Intron | NP_001309869.1 | |||
NM_001322941.1 | 4802 | Intron | NP_001309870.1 | |||
NM_001322943.1 | 4802 | Intron | NP_001309872.1 | |||
NM_001322944.1 | 4802 | Intron | NP_001309873.1 | |||
NM_013352.3 | 4802 | Intron | NP_037484.1 | |||
XM_017010795.1 | 4802 | Intron | XP_016866284.1 | |||
XM_017010796.1 | 4802 | Intron | XP_016866285.1 | |||
XM_017010797.1 | 4802 | Intron | XP_016866286.1 |
TSPYL1 - TSPY like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003309.3 | 4802 | UTR 3 | NP_003300.1 |
Set Membership: |
HapMap Validated |