Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCCCCATTCAGGTTCCCCAGAG[A/G]GTAGTGCCGAGGTGCTGACACCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604526 MIM: 614155 MIM: 614154 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IDH3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IDH3B - isocitrate dehydrogenase 3 (NAD(+)) beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258384.2 | Intron | NP_001245313.1 | ||||
NM_006899.4 | Intron | NP_008830.2 | ||||
NM_174855.3 | Intron | NP_777280.1 | ||||
XM_005260716.1 | Intron | XP_005260773.1 |
MIR1292 - microRNA 1292 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOP56 - NOP56 ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA51 - small nucleolar RNA, H/ACA box 51 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD110 - small nucleolar RNA, C/D box 110 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD56 - small nucleolar RNA, C/D box 56 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD57 - small nucleolar RNA, C/D box 57 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD86 - small nucleolar RNA, C/D box 86 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |