Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
74 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134580 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
F13B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
F13B - coagulation factor XIII B chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001994.2 | 1844 | Silent Mutation | AAC,AAT | N,N 602 | NP_001985.2 | |
XM_011509283.2 | 1844 | Silent Mutation | AAC,AAT | N,N 602 | XP_011507585.1 | |
XM_011509284.2 | 1844 | Silent Mutation | AAC,AAT | N,N 601 | XP_011507586.1 | |
XM_011509286.2 | 1844 | Silent Mutation | AAC,AAT | N,N 554 | XP_011507588.1 |