Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603859 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC25A13 - solute carrier family 25 member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160210.1 | Intron | NP_001153682.1 | ||||
NM_014251.2 | Intron | NP_055066.1 | ||||
XM_006715831.3 | Intron | XP_006715894.1 | ||||
XM_011515727.2 | Intron | XP_011514029.1 | ||||
XM_017011663.1 | Intron | XP_016867152.1 | ||||
XM_017011664.1 | Intron | XP_016867153.1 | ||||
XM_017011665.1 | Intron | XP_016867154.1 |
Set Membership: |
HapMap |