Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAAATGTCTCGGCTGTCCTCAG[A/G]CAAGGGGCATGCCCAAAAAGCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604890 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF3C5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GTF3C5 - general transcription factor IIIC subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122823.1 | Intron | NP_001116295.1 | ||||
NM_001286709.1 | Intron | NP_001273638.1 | ||||
NM_012087.3 | Intron | NP_036219.2 | ||||
XM_005272234.3 | Intron | XP_005272291.1 | ||||
XM_011519201.1 | Intron | XP_011517503.1 | ||||
XM_017015315.1 | Intron | XP_016870804.1 | ||||
XM_017015316.1 | Intron | XP_016870805.1 |
LOC105376306 - uncharacterized LOC105376306 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |