Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATCTTGGCTCCTAGAAACTTCT[G/T]CCCAGAGCCAGTCAGATCAGCCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612277 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAMTSL2 - ADAMTS like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145320.1 | Intron | NP_001138792.1 | ||||
NM_014694.3 | Intron | NP_055509.2 | ||||
XM_005272237.3 | Intron | XP_005272294.1 | ||||
XM_005272238.3 | Intron | XP_005272295.1 | ||||
XM_005272239.2 | Intron | XP_005272296.1 | ||||
XM_011519241.2 | Intron | XP_011517543.2 | ||||
XM_011519242.2 | Intron | XP_011517544.1 |
Set Membership: |
HapMap |