Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610823 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A42 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC25A42 - solute carrier family 25 member 42 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM161A - transmembrane protein 161A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256766.1 | 1666 | UTR 3 | NP_001243695.1 | |||
NM_017814.2 | 1666 | UTR 3 | NP_060284.1 | |||
XM_011528117.2 | 1666 | UTR 3 | XP_011526419.1 | |||
XM_017026916.1 | 1666 | UTR 3 | XP_016882405.1 |