Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601273 MIM: 190315 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLTCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLTCL1 - clathrin heavy chain like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001835.3 | Intron | NP_001826.3 | ||||
NM_007098.3 | Intron | NP_009029.3 | ||||
XM_011530401.2 | Intron | XP_011528703.1 | ||||
XM_017028953.1 | Intron | XP_016884442.1 | ||||
XM_017028954.1 | Intron | XP_016884443.1 | ||||
XM_017028955.1 | Intron | XP_016884444.1 | ||||
XM_017028956.1 | Intron | XP_016884445.1 | ||||
XM_017028957.1 | Intron | XP_016884446.1 |
SLC25A1 - solute carrier family 25 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |