Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTAAAACTTGTCAAATATAACAT[C/T]AAGTGAAAGTTAACATGAATTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606821 MIM: 601910 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COG5 - component of oligomeric golgi complex 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161520.1 | Intron | NP_001154992.1 | ||||
NM_006348.3 | Intron | NP_006339.3 | ||||
NM_181733.2 | Intron | NP_859422.2 |
GPR22 - G protein-coupled receptor 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005295.2 | Intron | NP_005286.2 | ||||
XM_011516056.2 | Intron | XP_011514358.1 |
Set Membership: |
HapMap |