Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610591 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ARHGAP27 - Rho GTPase activating protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282290.1 | 3571 | UTR 3 | NP_001269219.1 | |||
NM_174919.3 | 3571 | Intron | NP_777579.2 | |||
NM_199282.2 | 3571 | UTR 3 | NP_954976.1 | |||
XM_005257128.3 | 3571 | UTR 3 | XP_005257185.1 | |||
XM_006721745.2 | 3571 | UTR 3 | XP_006721808.1 | |||
XM_006721747.2 | 3571 | UTR 3 | XP_006721810.1 | |||
XM_006721750.3 | 3571 | Intron | XP_006721813.1 | |||
XM_011524468.1 | 3571 | UTR 3 | XP_011522770.1 | |||
XM_011524469.1 | 3571 | UTR 3 | XP_011522771.1 | |||
XM_011524470.2 | 3571 | UTR 3 | XP_011522772.1 | |||
XM_011524471.1 | 3571 | UTR 3 | XP_011522773.1 | |||
XM_011524472.2 | 3571 | UTR 3 | XP_011522774.1 | |||
XM_011524473.2 | 3571 | Intron | XP_011522775.1 | |||
XM_011524474.1 | 3571 | UTR 3 | XP_011522776.1 | |||
XM_017024311.1 | 3571 | UTR 3 | XP_016879800.1 | |||
XM_017024312.1 | 3571 | UTR 3 | XP_016879801.1 | |||
XM_017024313.1 | 3571 | Intron | XP_016879802.1 |
Set Membership: |
HapMap Validated |