Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610591 MIM: 611466 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGAP27 - Rho GTPase activating protein 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHM1 - pleckstrin homology and RUN domain containing M1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014798.2 | 4717 | UTR 3 | NP_055613.1 | |||
XM_006722201.3 | 4717 | UTR 3 | XP_006722264.1 | |||
XM_011525523.2 | 4717 | Intron | XP_011523825.1 | |||
XM_011525524.1 | 4717 | UTR 3 | XP_011523826.1 | |||
XM_011525525.1 | 4717 | UTR 3 | XP_011523827.1 | |||
XM_011525526.1 | 4717 | UTR 3 | XP_011523828.1 | |||
XM_011525528.2 | 4717 | UTR 3 | XP_011523830.1 | |||
XM_017025451.1 | 4717 | UTR 3 | XP_016880940.1 | |||
XM_017025452.1 | 4717 | UTR 3 | XP_016880941.1 | |||
XM_017025453.1 | 4717 | UTR 3 | XP_016880942.1 | |||
XM_017025454.1 | 4717 | UTR 3 | XP_016880943.1 |