Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACACAAATACTTGCCATTGTGTT[A/G]CAGTTGCCTATAGTATTTGGTACAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610679 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK14 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDK14 - cyclin dependent kinase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287135.1 | Intron | NP_001274064.1 | ||||
NM_001287136.1 | Intron | NP_001274065.1 | ||||
NM_001287137.1 | Intron | NP_001274066.1 | ||||
NM_012395.3 | Intron | NP_036527.1 | ||||
XM_005250436.3 | Intron | XP_005250493.1 | ||||
XM_005250438.3 | Intron | XP_005250495.1 | ||||
XM_005250439.2 | Intron | XP_005250496.1 | ||||
XM_011516306.2 | Intron | XP_011514608.1 | ||||
XM_011516307.2 | Intron | XP_011514609.1 | ||||
XM_017012320.1 | Intron | XP_016867809.1 | ||||
XM_017012321.1 | Intron | XP_016867810.1 | ||||
XM_017012322.1 | Intron | XP_016867811.1 |
Set Membership: |
HapMap |